2-(4-Bromophenyl)-4,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5797-1G B5797-200MG
TCB5797-1GEA 341 USD
TCB5797-1G TCB5797-200MG
2-(4-Bromophenyl)-4,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
2-(4-Bromophenyl)-4,6-diphenylpyrimidine
Formula: C₂₂H₁₅BrN₂
MW: 387.28 g/mol
MDL Number: MFCD29477690
CAS Number: 457613-56-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 233.0 to 238.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR