Cyclohexylamine Hydroiodide ≥95.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

Synonyms: Cyclohexylammonium iodide

C3532-1G C3532-5G
TCC3532-1GEA 41 USD
TCC3532-1G TCC3532-5G
Cyclohexylamine Hydroiodide ≥95.0% (by total nitrogen and titration analysis)
Cyclohexylamine Hydroiodide
Formula: C₆H₁₃N·HI
MW: 227.09 g/mol
Melting Pt: 194 °C
CAS Number: 45492-87-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR