1,4-Bis(dimethylvinylsilyl)benzene ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,4-Bis(dimethyl(vinyl)silyl)benzene, 1,4-Phenylenebis[dimethyl(vinyl)silane]

B6054-25G B6054-5G
TCB6054-25GEA 373 USD
TCB6054-25G TCB6054-5G
1,4-Bis(dimethylvinylsilyl)benzene ≥95.0% (by GC)
1,4-Bis(dimethylvinylsilyl)benzene
Formula: C₁₄H₂₂Si₂
MW: 246.50 g/mol
Boiling Pt: 95 °C
MDL Number: MFCD07778038
CAS Number: 4519-17-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR