1,4-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)

Supplier: TCI America
Warning

B5646-1G B5646-5G
TCB5646-1GEA 114 USD
TCB5646-1G TCB5646-5G
1,4-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)
1,4-Bis(trifluoromethyl)cyclohexane
Formula: C₈H₁₀F₆
MW: 220.16 g/mol
Boiling Pt: 140 °C
Melting Pt: –40 °C
MDL Number: MFCD29066795
CAS Number: 433-18-1
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR