2,2'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2'-Biphenyldiboronic acid bis(pinacol) ester

B5703-1G B5703-200MG
TCB5703-1GEA 274 USD
TCB5703-1G TCB5703-200MG
2,2'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl ≥98.0% (by GC, titration analysis)
2,2'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl
Formula: C₂₄H₃₂B₂O₄
MW: 406.14 g/mol
MDL Number: MFCD30729880
CAS Number: 398128-09-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR