(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97.0% (by GC)

Supplier: TCI America

Synonyms: (2R,5R)-5-Benzyl-2-(tert-butyl)-3-methylimidazolidin-4-one, (2R,5R)-(+)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone

B4137-1G B4137-200MG
TCB4137-1GEA 221 USD
TCB4137-1G TCB4137-200MG
(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97.0% (by GC)
(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone
Formula: C₁₅H₂₂N₂O
MW: 246.35 g/mol
MDL Number: MFCD08276842
CAS Number: 390766-89-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR