2-Acetoxypropionyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-Chloro-1-oxopropan-2-yl acetate, O-Acetyllactoyl chloride

A2932-1G A2932-5G
TCA2932-1GEA 157 USD
TCA2932-1G TCA2932-5G
2-Acetoxypropionyl chloride ≥97.0% (by GC, titration analysis)
2-Acetoxypropionyl chloride
Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18969061
CAS Number: 38939-83-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR