5-Amino-2,4,6-triiodoisophthaloyl Dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2854-25G A2854-5G
TCA2854-25GEA 543 USD
TCA2854-25G TCA2854-5G
5-Amino-2,4,6-triiodoisophthaloyl Dichloride ≥98.0% (by titrimetric analysis)
5-Amino-2,4,6-triiodoisophthaloyl Dichloride
Formula: C₈H₂Cl₂I₃NO₂
Melting Pt: 231 °C
MDL Number: MFCD00270836
CAS Number: 37441-29-5
EINECS: 417-220-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR