Chloro(hexyl)dimethylsilane ≥96.0% (by GC)

Supplier: TCI America
Danger

C3415-25ML C3415-5ML
TCC3415-25MLEA 205 USD
TCC3415-25ML TCC3415-5ML
Chloro(hexyl)dimethylsilane ≥96.0% (by GC)
Chloro(hexyl)dimethylsilane
Formula: C₈H₁₉ClSi
Boiling Pt: 184 °C
MDL Number: MFCD01073817
CAS Number: 3634-59-1
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR