2-Bromobenzaldehyde Dimethyl Acetal ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Bromo-2-(dimethoxymethyl)benzene

B5827-25G B5827-5G
TCB5827-25GEA 95 USD
TCB5827-25G TCB5827-5G
2-Bromobenzaldehyde Dimethyl Acetal ≥98.0% (by GC)
2-Bromobenzaldehyde Dimethyl Acetal
Formula: C₉H₁₁BrO₂
Boiling Pt: 103 °C
MDL Number: MFCD07780683
CAS Number: 35849-09-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR