2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC) stabilised

Supplier: TCI America

Synonyms: (3-Buten-1-yl)boronic acid Pinacol ester, 2-(But-3-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Stabilization: Stabilised with phenothiazine

B5960-1G B5960-5G
TCB5960-1GEA 131 USD
TCB5960-1G TCB5960-5G
2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC) stabilised
2-(3-Buten-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₀H₁₉BO₂
MDL Number: MFCD11858602
CAS Number: 331958-92-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR