2-Bromo-1,1':4',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

B5760-1G B5760-5G
TCB5760-1GEA 101 USD
TCB5760-1G TCB5760-5G
2-Bromo-1,1':4',1''-terphenyl ≥98.0% (by GC)
2-Bromo-1,1':4',1''-terphenyl
Formula: C₁₈H₁₃Br
MDL Number: MFCD00093613
CAS Number: 3282-24-4
UN: 3152
ADR: 9,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR