9-([1,1'-Biphenyl]-4-yl)anthracene ≥98.0% (by GC)
Supplier: TCI America
B5717-5G
B5717-1G
TCB5717-5GEA
176
USD
TCB5717-5G
TCB5717-1G
9-([1,1'-Biphenyl]-4-yl)anthracene ≥98.0% (by GC)
9-([1,1'-Biphenyl]-4-yl)anthracene
Formula:
C₂₆H₁₈ MW: 330.43 g/mol |
MDL Number:
MFCD27979543 CAS Number: 323195-31-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 221.0 to 225.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...