2-Benzoyl-5-bromothiophene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (5-Bromothiophen-2-yl)(phenyl)methanone

B5340-1G B5340-5G
TCB5340-1GEA 69 USD
TCB5340-1G TCB5340-5G
2-Benzoyl-5-bromothiophene ≥98.0% (by GC)
2-Benzoyl-5-bromothiophene
Formula: C₁₁H₇BrOS
MW: 267.14 g/mol
MDL Number: MFCD03086026
CAS Number: 31161-46-3

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR