2-(Chloromethyl)-6-methylpyridine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C3596-1G C3596-5G
TCC3596-1GEA 69 USD
TCC3596-1G TCC3596-5G
2-(Chloromethyl)-6-methylpyridine hydrochloride ≥98.0% (by titrimetric analysis)
2-(Chloromethyl)-6-methylpyridine hydrochloride
Formula: C₇H₈ClN·HCl
MW: 178.06 g/mol
MDL Number: MFCD01691969
CAS Number: 3099-30-7
EINECS: 221-451-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR