2-tert-Butyl-1,1,3,3-tetramethylguanidine ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B6052-1G B6052-5G
TCB6052-1GEA 50 USD
TCB6052-1G TCB6052-5G
2-tert-Butyl-1,1,3,3-tetramethylguanidine ≥95.0% (by GC, titration analysis)
2-tert-Butyl-1,1,3,3-tetramethylguanidine
Formula: C₉H₂₁N₃
Flash Pt: 65 °C
MDL Number: MFCD01862972
CAS Number: 29166-72-1
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR