2-Chloro-10-(3-chloropropyl)-10H-phenothiazine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3394-1G C3394-250MG
TCC3394-1GEA 677 USD
TCC3394-1G TCC3394-250MG
2-Chloro-10-(3-chloropropyl)-10H-phenothiazine ≥98.0% (by GC)
2-Chloro-10-(3-chloropropyl)-10H-phenothiazine
Formula: C₁₅H₁₃Cl₂NS
Boiling Pt: 135 °C
MDL Number: MFCD01247123
CAS Number: 2765-59-5
EINECS: 220-438-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR