Benzyl Benzodithioate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Benzyl benzodithioate, Benzyl Benzenecarbodithioate, Benzodithioic acid Benzyl ester, Benzenecarbodithioic acid Benzyl ester

B6066-1G B6066-5G
TCB6066-1GEA 181 USD
TCB6066-1G TCB6066-5G
Benzyl Benzodithioate ≥95.0% (by GC)
Benzyl Benzodithioate
Formula: C₁₄H₁₂S₂
MW: 244.37 g/mol
Boiling Pt: 254 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08705657
CAS Number: 27249-90-7

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR