Butyltrimethylammonium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: N,N,N-Trimethylbutan-1-aminium bromide

B5798-25G B5798-5G
TCB5798-25GEA 218 USD
TCB5798-25G TCB5798-5G
Butyltrimethylammonium bromide ≥97.0% (by titrimetric analysis)
Butyltrimethylammonium bromide
Formula: C₇H₁₈BrN
MW: 196.13 g/mol
Melting Pt: 196 °C
MDL Number: MFCD12761450
CAS Number: 2650-51-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR