Cinepazide Maleate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-[(1-Pyrrolidinylcarbonyl)methyl]-4-(3,4,5-trimethoxycinnamoyl)piperazine Maleate, Vasodistal, 1-(4-(2-Oxo-2-(pyrrolidin-1-yl)ethyl)piperazin-1-yl)-3-(3,4,5-trimethoxyphenyl)prop-2-en-1-one maleate

C3474-250MG C3474-25MG C3474-1G
TCC3474-250MGEA 24 USD
TCC3474-250MG TCC3474-25MG TCC3474-1G
Cinepazide Maleate ≥98.0% (by HPLC, titration analysis)
Cinepazide Maleate
Formula: C₂₂H₃₁N₃O₅·C₄H₄O₄
MW: 533.58 g/mol
MDL Number: MFCD01320526
CAS Number: 26328-04-1
EINECS: 247-613-1
Merck Index: 14,02291

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR