4,4''-Dibromo-1,1':2',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

D4998-1G D4998-200MG
TCD4998-1GEA 342 USD
TCD4998-1G TCD4998-200MG
4,4''-Dibromo-1,1':2',1''-terphenyl ≥98.0% (by GC)
4,4''-Dibromo-1,1':2',1''-terphenyl
Formula: C₁₈H₁₂Br₂
MDL Number: MFCD28138092
CAS Number: 24253-43-8
UN: 3152
ADR: 9,II

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR