tert-Butyl Hydrogen Dodecanedioate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 12-(tert-Butoxy)-12-oxododecanoic acid, Dodecanedioic acid Mono-tert-butyl ester

B5926-1G B5926-5G
TCB5926-1GEA 203 USD
TCB5926-1G TCB5926-5G
tert-Butyl Hydrogen Dodecanedioate ≥98.0% (by GC, titration analysis)
tert-Butyl Hydrogen Dodecanedioate
Formula: C₁₆H₃₀O₄
Storage Temperature: Refrigerator
MDL Number: MFCD28098251
CAS Number: 234081-98-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR