2,2'-Bipyridine-5,5'-diol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5,5'-Dihydroxy-2,2'-bipyridyl, 5,5'-Dihydroxy-2,2'-bipyridine

B5525-250MG B5525-50MG
TCB5525-250MGEA 407 USD
TCB5525-250MG TCB5525-50MG
2,2'-Bipyridine-5,5'-diol ≥98.0% (by GC, titration analysis)
2,2'-Bipyridine-5,5'-diol
Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 276 °C
MDL Number: MFCD20488016
CAS Number: 2326-78-5

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR