4-Bromo-4'-chloro-1,1'-biphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromo-4'-Chloro-1,1'-biphenyl

B5966-1G B5966-5G
TCB5966-1GEA 37 USD
TCB5966-1G TCB5966-5G
4-Bromo-4'-chloro-1,1'-biphenyl ≥98.0% (by GC)
4-Bromo-4'-chloro-1,1'-biphenyl
Formula: C₁₂H₈BrCl
MW: 267.55 g/mol
MDL Number: MFCD06201384
CAS Number: 23055-77-8
UN: 3152
ADR: 9,II

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR