(E)-4-(Dimethylamino)chalcone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D5312-1G
D5312-200MG
TCD5312-1GEA
346
USD
TCD5312-1G
TCD5312-200MG
(E)-4-(Dimethylamino)chalcone ≥98.0% (by GC, titration analysis)
(E)-4-(Dimethylamino)chalcone
Formula:
C₁₇H₁₇NO |
CAS Number:
22965-98-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 112.0 to 116.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...