4-Bromo-4'-(4-bromophenyl)-3',5',6'-triphenyl-1,1':2',1''-terphenyl ≥98.0% (by HPLC)

Supplier: TCI America

B5986-1G B5986-5G
TCB5986-1GEA 130 USD
TCB5986-1G TCB5986-5G
4-Bromo-4'-(4-bromophenyl)-3',5',6'-triphenyl-1,1':2',1''-terphenyl ≥98.0% (by HPLC)
4-Bromo-4'-(4-bromophenyl)-3',5',6'-triphenyl-1,1':2',1''-terphenyl
Formula: C₄₂H₂₈Br₂
Melting Pt: 374 °C
MDL Number: MFCD32062866
CAS Number: 22932-54-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR