2-Bromo-4,5-dimethylphenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B5394-1G B5394-5G
TCB5394-1GEA 34 USD
TCB5394-1G TCB5394-5G
2-Bromo-4,5-dimethylphenol ≥98.0% (by GC, titration analysis)
2-Bromo-4,5-dimethylphenol
Formula: C₈H₉BrO
MDL Number: MFCD09260834
CAS Number: 22802-39-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR