6-Bromo-7-hydroxy-4-(hydroxymethyl)coumarin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Bromo-7-hydroxy-4-(hydroxymethyl)-2H-chromen-2-one

B5738-1G B5738-250MG
TCB5738-1GEA 792 USD
TCB5738-1G TCB5738-250MG
6-Bromo-7-hydroxy-4-(hydroxymethyl)coumarin ≥98.0% (by titrimetric analysis)
6-Bromo-7-hydroxy-4-(hydroxymethyl)coumarin
Formula: C₁₀H₇BrO₄
MW: 271.07 g/mol
MDL Number: MFCD25961202
CAS Number: 223420-41-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR