N,N-Bis(4-bromophenyl)-[1,1':4',1''-terphenyl]-4-amine ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
B6126-1G
B6126-5G
TCB6126-1GEA
141
USD
TCB6126-1G
TCB6126-5G
N,N-Bis(4-bromophenyl)-[1,1':4',1''-terphenyl]-4-amine ≥97.0% (by HPLC, total nitrogen)
N,N-Bis(4-bromophenyl)-[1,1':4',1''-terphenyl]-4-amine
Formula:
C₃₀H₂₁Br₂N |
CAS Number:
2205054-16-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 193.0 to 197.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...