2-Chloro-4,6-di-p-tolyl-1,3,5-triazine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Chloro-4,6-bis(4-methylphenyl)-1,3,5-triazine

C3514-1G C3514-5G
TCC3514-1GEA 110 USD
TCC3514-1G TCC3514-5G
2-Chloro-4,6-di-p-tolyl-1,3,5-triazine ≥98.0% (by GC)
2-Chloro-4,6-di-p-tolyl-1,3,5-triazine
Formula: C₁₇H₁₄ClN₃
MW: 295.77 g/mol
MDL Number: MFCD29071841
CAS Number: 21902-34-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 205.0 to 209.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR