2-(2H-Benzotriazol-2-yl)-4-methyl-6-(2-propenyl)phenol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Allyl-6-(2H-benzotriazol-2-yl)-4-methylphenol

B5709-1G B5709-5G
TCB5709-1GEA 156 USD
TCB5709-1G TCB5709-5G
2-(2H-Benzotriazol-2-yl)-4-methyl-6-(2-propenyl)phenol ≥98.0% (by GC)
2-(2H-Benzotriazol-2-yl)-4-methyl-6-(2-propenyl)phenol
Formula: C₁₆H₁₅N₃O
MW: 265.32 g/mol
MDL Number: MFCD00236141
CAS Number: 2170-39-0

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR