4-Bromo-1,2-diiodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5743-1G B5743-5G
TCB5743-1GEA 101 USD
TCB5743-1G TCB5743-5G
4-Bromo-1,2-diiodobenzene ≥98.0% (by GC)
4-Bromo-1,2-diiodobenzene
Formula: C₆H₃BrI₂
MDL Number: MFCD07778994
CAS Number: 21521-54-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR