2-(4-Biphenylyl)indole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-([1,1'-Biphenyl]-4-yl)-1H-indole

B5250-1G B5250-5G
TCB5250-1GEA 140 USD
TCB5250-1G TCB5250-5G
2-(4-Biphenylyl)indole ≥98.0% (by GC)
2-(4-Biphenylyl)indole
Formula: C₂₀H₁₅N
MW: 269.35 g/mol
MDL Number: MFCD00417190
CAS Number: 21470-37-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR