9-Aminoanthracene hydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Anthracen-9-amine hydrochloride

A3208-1G A3208-5G
TCA3208-1GEA 208 USD
TCA3208-1G TCA3208-5G
9-Aminoanthracene hydrochloride ≥98.0% (by total nitrogen basis)
9-Aminoanthracene hydrochloride
Formula: C₁₄H₁₁N·HCl
MDL Number: MFCD00050528
CAS Number: 209743-32-8

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR