beta-Alanine Hydroiodide, low water content ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

Synonyms: H-beta-Ala-OH.HI, 3-Aminopropionic acid Hydroiodide

A3112-1G A3112-5G
TCA3112-1GEA 85 USD
TCA3112-1G TCA3112-5G
beta-Alanine Hydroiodide, low water content ≥98.0% (by total nitrogen and titration analysis)
beta-Alanine Hydroiodide
Formula: C₃H₇NO₂·HI
MW: 217.01 g/mol
Melting Pt: 199 °C
CAS Number: 2096495-59-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR