(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

C3202-25G C3202-5G
TCC3202-25GEA 308 USD
TCC3202-25G TCC3202-5G
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile ≥98.0% (by GC)
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile
Formula: C₇H₉ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD08689902
CAS Number: 207557-35-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C
Specific rotation [a]20/D -150.0 to -160.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR