Benzo[b]naphtho[1,2-d]furan ≥98.0% (by GC)

Supplier: TCI America
Warning

B5884-1G B5884-5G
TCB5884-1GEA 108 USD
TCB5884-1G TCB5884-5G
Benzo[b]naphtho[1,2-d]furan ≥98.0% (by GC)
Benzo[b]naphtho[1,2-d]furan
Formula: C₁₆H₁₀O
MDL Number: MFCD00046916
CAS Number: 205-39-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR