Acrylic anhydride ≥95.0% (by GC, titration analysis) stabilised
Supplier: TCI America
|
Danger
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A3243-1G
A3243-5G
TCA3243-1GEA
24
USD
TCA3243-1G
TCA3243-5G
Acrylic anhydride ≥95.0% (by GC, titration analysis) stabilised
Acrylic anhydride
Formula:
C₆H₆O₃ Boiling Pt: 67 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00048146 CAS Number: 2051-76-5 EINECS: 218-128-2 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Morpholine Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...