9,9'-Diethyl-9H,9'H-3,3'-bicarbazole ≥97.0% (by total nitrogen basis)

Supplier: TCI America
Warning

D5387-1G D5387-5G
TCD5387-1GEA 124 USD
TCD5387-1G TCD5387-5G
9,9'-Diethyl-9H,9'H-3,3'-bicarbazole ≥97.0% (by total nitrogen basis)
9,9'-Diethyl-9H,9'H-3,3'-bicarbazole
Formula: C₂₈H₂₄N₂
MW: 388.51 g/mol
MDL Number: MFCD32062883
CAS Number: 20466-00-6

Order Now

Specification Test Results

Appearance White to Amber powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 190.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR