AlkylFluor(TM) ≥97.0% (by total nitrogen basis)

Supplier: TCI America
Danger

B5480-1G B5480-200MG
TCB5480-1GEA 441 USD
TCB5480-1G TCB5480-200MG
AlkylFluor(TM) ≥97.0% (by total nitrogen basis)
AlkylFluor(TM)
Formula: C₂₇H₃₆BF₅N₂
CAS Number: 2043361-32-4
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR