2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone ≥98.0% (by GC)

Supplier: TCI America

B6031-1G B6031-5G
TCB6031-1GEA 43 USD
TCB6031-1G TCB6031-5G
2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone ≥98.0% (by GC)
2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone
Formula: C₁₁H₁₀BrFO
MW: 257.10 g/mol
Boiling Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977644
CAS Number: 204205-33-4

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR