Bilastine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-(2-(4-(1-(2-Ethoxyethyl)-1H-benzo[d]imidazol-2-yl)piperidin-1-yl)ethyl)phenyl)-2-methylpropanoic acid, 2-[4-[2-[4-[1-(2-Ethoxyethyl)-1H-benzo[d]imidazol-2-yl]piperidin-1-yl]ethyl]phenyl]-2-methylpropanoic acid

B5392-1G B5392-250MG
TCB5392-1GEA 258 USD
TCB5392-1G TCB5392-250MG
Bilastine ≥98.0% (by HPLC, titration analysis)
Bilastine
Formula: C₂₈H₃₇N₃O₃
MW: 463.62 g/mol
MDL Number: MFCD09837814
CAS Number: 202189-78-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 200.0 to 204.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR