N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N1,N1'-([1,1'-Biphenyl]-4,4'-diyl)bis(N1-phenyl-N4,N4-di-m-tolylbenzene-1,4-diamine), DNTPD

B5634-5G B5634-1G
TCB5634-5GEA 523 USD
TCB5634-5G TCB5634-1G
N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine
Formula: C₆₄H₅₄N₄
MW: 879.16 g/mol
MDL Number: MFCD22372686
CAS Number: 199121-98-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR