2-(4-Chlorophenyl)-2-propanol ≥98.0% (by GC)

Supplier: TCI America

C3324-1G C3324-5G
TCC3324-1GEA 110 USD
TCC3324-1G TCC3324-5G
2-(4-Chlorophenyl)-2-propanol ≥98.0% (by GC)
2-(4-Chlorophenyl)-2-propanol
Formula: C₉H₁₁ClO
Boiling Pt: 96 °C
MDL Number: MFCD11870102
CAS Number: 1989-25-9
EINECS: 217-865-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR