4,7-Dihydroxycoumarin ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4,7-Dihydroxy-2H-chromen-2-one

D5175-1G D5175-5G
TCD5175-1GEA 95 USD
TCD5175-1G TCD5175-5G
4,7-Dihydroxycoumarin ≥98.0% (by HPLC)
4,7-Dihydroxycoumarin
Formula: C₉H₆O₄
MW: 178.14 g/mol
MDL Number: MFCD00203882
CAS Number: 1983-81-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR