3,5-Di-tert-butylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3,5-Di-tert-butylbenzeneboronic acid, 3,5-bis(tert-butyl)phenylboronic acid, (3,5-Di-tert-butylphenyl)boronic acid

D5072-200MG D5072-500MG
TCD5072-200MGEA 88 USD
TCD5072-200MG TCD5072-500MG
3,5-Di-tert-butylphenylboronic acid (contains varying amounts of Anhydride)
3,5-Di-tert-butylphenylboronic acid
Formula: C₁₄H₂₃BO₂
MW: 234.15 g/mol
Melting Pt: 182 °C
MDL Number: MFCD07780212
CAS Number: 197223-39-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR