5-Bromobenzo[b]naphtho[1,2-d]thiophene ≥97.0% (by GC)

Supplier: TCI America

B5940-1G B5940-5G
TCB5940-1GEA 152 USD
TCB5940-1G TCB5940-5G
5-Bromobenzo[b]naphtho[1,2-d]thiophene ≥97.0% (by GC)
5-Bromobenzo[b]naphtho[1,2-d]thiophene
Formula: C₁₆H₉BrS
MW: 313.21 g/mol
CAS Number: 189097-35-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR