2-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

B6000-1G B6000-200MG
TCB6000-1GEA 244 USD
TCB6000-1G TCB6000-200MG
2-Bromo-5'-phenyl-1,1':3',1''-terphenyl ≥98.0% (by GC)
2-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formula: C₂₄H₁₇Br
MDL Number: MFCD32062871
CAS Number: 1890136-54-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR