1,3-Bis[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]-1,1,3,3-tetramethyldisiloxane ≥95.0% (by GC)

Supplier: TCI America
Warning

B5922-25G B5922-5G
TCB5922-25GEA 172 USD
TCB5922-25G TCB5922-5G
1,3-Bis[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]-1,1,3,3-tetramethyldisiloxane ≥95.0% (by GC)
1,3-Bis[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]-1,1,3,3-tetramethyldisiloxane
Formula: C₂₀H₃₈O₃Si₂
Boiling Pt: 195 °C
Flash Pt: 200 °C
MDL Number: MFCD03262840
CAS Number: 18724-32-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR