3-Chloropyridine N-Oxide ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Chloropyridine 1-oxide

C2329-1G C2329-5G
TCC2329-1GEA 143 USD
TCC2329-1G TCC2329-5G
3-Chloropyridine N-Oxide ≥98.0% (by GC, titration analysis)
3-Chloropyridine N-Oxide
Formula: C₅H₄ClNO
MW: 129.54 g/mol
Boiling Pt: 89 °C
MDL Number: MFCD00128857
CAS Number: 1851-22-5

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR